Zinc ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS
Danger

30-0050-25G 30-0050-5G
100209-898EA 561.91 USD
100209-898 100209-900
Zinc ≥99.999%, powder
Zinc
Formula: Zn
Boiling Pt: 907 °C
Melting Pt: 419.5 °C
Density: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR