Copper ≥99%, powder

Supplier: STREM CHEMICALS INC MS
Danger

29-0050-500G 29-0050-2000G
100205-082EA 84.92 USD
100205-082 100207-648
Copper ≥99%, powder
Copper
Formula: Cu
Boiling Pt: 2567 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8,92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR