Nickel ≥99.995%, wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
28-1500-25CM
28-1500-100CM
100198-736EA
186.83
USD
100198-736
100206-556
Nickel ≥99.995%, wire
Nickel
Formula:
Ni Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8.90 Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III Merck Index: 12,06582 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...