Tetrabutylammonium tetrahydroborate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tetra-n-butylammonium borohydride
, Tetrabutylammonium borohydride
, Tetrabutylammonium tetrahydridoborate
02-5000-10G
02-5000-50G
100197-284EA
287.32
USD
100197-284
100197-984
Tetrabutylammonium tetrahydroborate ≥97%
Tetrabutylammonium tetrahydroborate
Formula:
C₁₆H₄₀BN MW: 257.31 g/mol Melting Pt: 124…128 °C |
MDL Number:
MFCD00012035 CAS Number: 33725-74-5 EINECS: 251-658-2 UN: 2813 ADR: 4.3,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...