Chromium(III) chloride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chromic chloride, Chromium trichloride

24-0270-50G 24-0270-250G
100200-654EA 124.55 USD
100200-654 100210-802
Chromium(III) chloride, anhydrous ≥99%
Chromium (III) chloride
Formula: Cl₃Cr
MW: 158.35 g/mol
Boiling Pt: 1300 °C (subl.)
Melting Pt: 1152
Density: 2.76
Storage Temperature: Ambient
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 12,02278

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR