Calcium hydrogen orthophosphate ≥99.95% (Ca basis)

Supplier: STREM CHEMICALS INC MS

20-2035-250G 20-2035-50G
100201-516EA 397.72 USD
100201-516 100204-006
Calcium hydrogen orthophosphate ≥99.95% (Ca basis)
Calcium hydrogen orthophosphate
Formula: CaHPO₄
Melting Pt: 370 °C
Density: 2.89 g/cm³ (20 °C)
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR