Calcium oxide ≥99.95% (Ca basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lime
, Quicklime
, caustic
, Calcium oxide from marble
20-2030-10G
20-2030-50G
100207-614EA
43.88
USD
100207-614
100200-886
Calcium oxide ≥99.95% (Ca basis)
Calcium oxide
Formula:
CaO MW: 56.08 g/mol Boiling Pt: 2850 °C Melting Pt: 2580 °C Density: 3.30 Storage Temperature: Ambient |
MDL Number:
MFCD00010911 CAS Number: 1305-78-8 EINECS: 215-138-9 UN: 1910 ADR: 8,III Merck Index: 13,01691 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...