Calcium carbonate ≥99.999% (Ca basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

20-0200-50G 20-0200-10G
100201-196EA 549.17 USD
100201-196 100196-134
Calcium carbonate ≥99.999% (Ca basis), PURATREM
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2.930
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR