Potassium hydroxide ≥85%, pellets ACS

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

19-3150-2000G 19-3150-500G
100204-526EA 185.42 USD
100204-526 100208-368
Potassium hydroxide ≥85%, pellets ACS
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 2.044
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR