Triphenylphosphine ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-6700-100G
15-6700-500G
15-6700-2000G
100207-592EA
42.46
USD
100207-592
100207-594
100196-112
Triphenylphosphine ≥99%
Triphenylphosphine
Phosphine
Formula:
C₁₈H₁₅P MW: 262.29 g/mol Boiling Pt: 377 Melting Pt: 79 Density: 1.194 g/cm³ (20 °C) Flash Pt: 359 °F Storage Temperature: Ambient |
MDL Number:
MFCD00003043 CAS Number: 603-35-0 EINECS: 210-036-0 Merck Index: 12,09873 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...