Triethylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger


Packed in a Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC).

15-6302-25G
100198-136EA 510.95 USD
100198-136
Triethylphosphine ≥99%
Triethylphosphine

Phosphine

Formula: C₆H₁₅P
Boiling Pt: 126…128 °C
Melting Pt: –3 °C
Density: 0.81
Flash Pt: 1 °F
MDL Number: MFCD00009256
CAS Number: 554-70-1
EINECS: 209-068-8
UN: 2845
ADR: 4.2,I
Merck Index: 12,09807

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR