Triethylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

15-6300-25G 15-6300-5G
100201-496EA 103.32 USD
100201-496 100201-494
Triethylphosphine ≥99%
Triethylphosphine

Phosphine

Formula: C₆H₁₅P
Boiling Pt: 126…128 °C
Melting Pt: –3 °C
Density: 0.81
Flash Pt: 1 °F
MDL Number: MFCD00009256
CAS Number: 554-70-1
EINECS: 209-068-8
UN: 2845
ADR: 4.2,I
Merck Index: 12,09807

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR