Magnesium oxide ≥99.99% (Mg basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Magnesia usta
12-2025-5G
12-2025-25G
100203-958EA
106.15
USD
100203-958
100201-428
Magnesium oxide ≥99.99% (Mg basis), PURATREM
Magnesium oxide
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 Melting Pt: 2800 Density: 3.58 |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...