Anti-IgG gamma Goat Polyclonal Antibody

Supplier: SeraCare
5210-0171
95058-000EA 396.31 USD
95058-000
Anti-IgG gamma Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG (g) Reacts with Pig (Swine, Porcine)

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR