Anti-Salmonella Mol. Grade Goat Polyclonal Antibody

Supplier: SeraCare
5310-0323
95058-078EA 975.58 USD
95058-078
Anti-Salmonella Mol. Grade Goat Polyclonal Antibody
Antibodies
Primary Goat Anti-Salmonella Mol. Grade Reacts with Salmonella

Type: Primary
Antigen: Salmonella Mol. Grade
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Goat
Isotype:
Reactivity: Salmonella
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR