Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: SeraCare
5220-0335
95058-732EA 906 USD
95058-732
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Rabbit (Lapine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR