Anti-IgM µ Goat Polyclonal Antibody (Biotin)

Supplier: SeraCare
5260-0041
95058-868EA 178.04 USD
95058-868
Anti-IgM µ Goat Polyclonal Antibody (Biotin)
Antibodies
Secondary Goat Anti-IgM (µ) Reacts with Rat

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR