Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: KPL
5220-0336 5450-0010 5220-0283
95058-730EA 194.19 USD
95058-730 95059-086 95058-230
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Rabbit (Lapine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

SPECIFICATIONS

Antigen Symbol IgG IgG
Environmentally Preferable
Antigen Name Immunoglobulin G Immunoglobulin G
Antibody Type Secondary Secondary
Clonality Polyclonal Polyclonal
Conjugation HRP (Horseradish Peroxidase) HRP (Horseradish Peroxidase)
Reactivity Rabbit Rabbit
Host Goat Goat
Isotype
Western Blot Yes Yes
ELISA Yes Yes
ImmunoChemistry Yes Yes
ImmunoPrecipitation Yes Yes
Flow Cytometry Yes Yes
Epitope Heavy and Light chain

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR