Anti-IgG Goat Polyclonal Antibody (DyLight® 488)

Supplier: SeraCare
5230-0385
95058-500EA 380.34 USD
95058-500
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Rabbit (Lapine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR