Anti-IgG Goat Polyclonal Antibody (Cy3®)

Supplier: SeraCare
5230-0366
95058-440EA 380.13 USD
95058-440
Anti-IgG Goat Polyclonal Antibody (Cy3®)
Antibodies
Secondary Goat Anti-IgG(H+L)RbSA Reacts with Mouse

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR