Anti-IgA alpha Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: SeraCare
5220-0367
95058-788EA 304.91 USD
95058-788
Anti-IgA alpha Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgA (α) Reacts with Mouse

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: alpha chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR