Anti-IgM µ Goat Polyclonal Antibody

Supplier: SeraCare
5210-0157
95057-991EA 354.37 USD
95057-991
Anti-IgM µ Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgM (mu) Reacts with Human

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR