Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: SeraCare
5220-0328 5220-0457
95058-716EA 419.38 USD
95058-716 95059-082
Anti-IgM µ Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgM (mu) Reacts with Human

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR