Anti-IgG Goat Polyclonal Antibody

Supplier: SeraCare
5210-0159
95057-993EA 135.11 USD
95057-993
Anti-IgG Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Human

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR