Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: SeraCare
5220-0330 5450-0009 5220-0277
95058-218EA 107.24 USD
95058-218 95059-084 95058-720
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Human

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR