Anti-IgG gamma Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: KPL
5220-0327 5220-0456
95058-714EA 451.66 USD
95058-714 95059-080
Anti-IgG gamma Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Secondary Goat Anti-IgG (gamma) Reacts with Human

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR