Anti-IgM µ Goat Polyclonal Antibody (DyLight® 488)

Supplier: SeraCare
5230-0381
95058-492EA 765.92 USD
95058-492
Anti-IgM µ Goat Polyclonal Antibody (DyLight® 488)
Antibodies
Secondary Goat Anti-IgM (mu) Reacts with Human

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR