Anti-IgA alpha Goat Polyclonal Antibody (Biotin)

Supplier: SeraCare
5260-0027
95058-840EA 184.92 USD
95058-840
Anti-IgA alpha Goat Polyclonal Antibody (Biotin)
Antibodies
Secondary Goat Anti-IgA (α) Reacts with Human

Type: Secondary
Antigen: IgA alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: alpha chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR