Anti-IgG Goat Polyclonal Antibody

Supplier: SeraCare
5210-0201
95058-052EA 143.19 USD
95058-052
Anti-IgG Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Horse (Equine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR