Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)

Supplier: SeraCare
5230-0383
95058-496EA 429.08 USD
95058-496
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 488)
Antibodies
Secondary Rabbit Anti-IgG (H+L) Reacts with Goat

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR