Anti-O157:H7 Goat Polyclonal Antibody

Supplier: SeraCare
5310-0327
95058-086EA 745.14 USD
95058-086
Anti-O157:H7 Goat Polyclonal Antibody
Antibodies
Primary Goat Anti-E.coli O157:H7 Mol. Grade Reacts with E.coli

Type: Primary
Antigen: O157:H7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR