Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: SeraCare
5220-0387
95058-818EA 408.91 USD
95058-818
Anti-IgG Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Secondary Goat Anti-IgG(H+L) Reacts with Dog (Canine)

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR