Anti-Borrelia species Antibody

Supplier: SeraCare
5310-0338
95058-092EA 633.69 USD
95058-092
Anti-Borrelia species Antibody
Antibodies
Primary Goat Anti-Borrelia species Reacts with Borrelia

Type: Primary
Antigen: Borrelia species
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Goat
Isotype:
Reactivity: Borrelia
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR