Utility Grade Masking Tapes, ORS Nasco, INC.
Supplier: ORS Nasco
761-87202 761-87217
300074-272CS 186.6 USD
300074-272 300074-270
Utility Grade Masking Tapes, ORS Nasco, INC.
Tapes
This utility grade masking tape has an aggressive adhesive designed for applications such as textile, carpet seaming, light industrial, manufacturing, bundling and tabbing.