High Tension Hacksaws, Essendant
Supplier: Stanley
680-15-113
300003-924EA 81.96 USD
300003-924
High Tension Hacksaws, Essendant
Saws
Fully adjustable tension-holds blades up to 32,000 psi.
  • Blades extended out nose of frame can be locked into place for tight clearance work
  • Quick-release blade-changing mechanism in handle
  • Tension guide
  • Blade storage in frame