Cryogenic Transfer Hoses, Western Enterprises
Supplier: Safety & Industrial Supplies
312-WMH-2-14 312-WMH-2-16 312-WMH-2-17 312-WMH-2-18
300067-120EA 815.86 USD
300067-120 300008-025 300067-122 300067-124
Cryogenic Transfer Hoses, Western Enterprises
Welding Supplies
  • Cleaned for oxygen service
  • Exceptional flexibility, hose resists kinking
  • Machined CGA stainless steel end connections
  • Constructed of 316 stainless steel for all wetted parts
  • Use in cyro-biological and medical systems, and gas filling plants