RS Series Medium Duty Single Stage Regulators, Western Enterprises
Supplier: Western Enterprises
RS Series
312-RS-7-4 312-RS-1-1 312-RS-7-5
300066-966EA 326.02 USD
300066-966 300066-976 300066-978
RS Series Medium Duty Single Stage Regulators, Western Enterprises
Welding Supplies
  • 450 PSIG models have KEL-F® seat accommodate high pressure applications
  • Dual inlet filter system extends regulator life
  • Hydrogen and methane models equipped with pipe away relief valves
  • Nickel chrome plated zinc bonnet