Flowmeters/Regulators, Gentec
Supplier: Gentec
331-191AR-60
300008-122EA 214.71 USD
300008-122
Flowmeters/Regulators, Gentec
Welding Supplies
  • Underwriters Laboratories Inc. listed
  • Single stage construction
  • Sintered bronze inlet filter
  • Chrome plated housing cap
  • Easily read high-pressure gauge
  • Internal safety pressure relief design
  • Calibrated for argon, helium and carbon dioxide gases
  • Polycarbonate flow tube and outer cover are impact resistant for maximum durability and service life
  • 0-70 SCFH Argon flow, 0-150 SCFH Helium, 0-60 SCFH Carbon Dioxide flow
  • Polycarbonate flow tube and outer cover are impact resistant for longer life
  • Flowmeter is back pressure compensated to ensure accurate reading when line restrictions are present
  • 2" high pressure gauge for easy reading