Safety Barricade Tapes, Empire® Level, ORS Nasco, INC.
Supplier: Safety & Industrial Supplies
272-11-081 272-77-0201
300007-629EA 24 USD
300007-629 300007-665
Safety Barricade Tapes, Empire® Level, ORS Nasco, INC.
Tapes
Reusable safety tape with heavy-duty 300 lbs. break strength is 16× stronger than regular barricade tape.
Reinforced fibres run throughout the tape.