Comfo® Respirator Cartridges, MSA
Supplier: Safety & Industrial Supplies
454-464032 454-464031 454-464033 454-815178 454-815177 454-815182 454-464046 454-492790 454-815180
300081-056PK 145.42 USD
300081-056 300081-138 300081-054 300081-058 300081-140 300052-940 300052-942 300081-060 300081-092 300081-142
Comfo® Respirator Cartridges, MSA
Respirators Reusable Respirators Respirator Filters
  • N95-Particulate filter (95% filter efficiency level) effective against particulate aerosols free of oil
  • R95-Particulate filter (95% filter efficiency level) effective against all particulate aerosols
  • P100-Particulate filter (99.97% filter efficiency level) effective against all particulate aerosols