Eye Wash & Shower Stations, Guardian
Supplier: Guardian
333-G1902 333-G1902HFC 333-G1902P
300068-326EA 1386.53 USD
300068-326 300068-328 300068-330
Eye Wash & Shower Stations, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Eye wash is activated by flag handle, two GS-Plus™ spray-type outlet heads deliver water for rinsing eyes.
  • Eye Wash Valve: 1/2" IPS chrome-plated brass stay-open ball valve
  • Shower Valve: 1" IPS chrome-plated brass stay-open ball valve, furnished with stainless steel actuating arm and 29" stainless steel pull rod.
  • Spray head assembly: two GS-Plus™ spray heads each have 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow.
  • Pipe and fittings: schedule 40 galvanized steel, furnished with orange polyethylene covers for vertical piping for high visibility and corrosion resistance.
Bowl height: 36", bowl dia. 11.5" or 12"
Outlet: 1.25"
Shower head Safety Orange ABS dia. 10x85"