DBI-SALA® Delta™ II No-Tangle™ Construction Harness
Supplier: Safety & Industrial Supplies
098-1102526 098-1103512
300075-528EA 250.86 USD
300075-528 300075-538 300075-540
DBI-SALA® Delta™ II No-Tangle™ Construction Harness
Fall Protection Fall Harnesses
Delta's No-Tangle™ design ensures comfort and simple donning. Adjustable non-slip chest strap with convenient pass-thru buckle.
  • Parachute buckles on lower shoulder straps for quick and easy adjustment
  • Spring loaded stand-up back D-ring ensures fast, easy, and safe connections
  • Built-in belt loops to support a SALA belt or one of your own
  • Wrap around no-tangle design slips on like a vest, making it quicker and easier to put on while providing ventilation, comfort, padding and security