Oil-Free Hand Carry Compressors, DeWalt®
Supplier: DeWalt
115-D55140
300060-396EA 402.25 USD
300060-396
Oil-Free Hand Carry Compressors, DeWalt®
Pumps Compressors
Flow rate: 0.60 CFM @ 100 psi
  • Durable oil free pump provides extended maintenance-free operation
  • Roll cage and control panel provide protection to key compressor components
  • Low amp draw motor provides easy start-up and reduces breaker tripping
  • Ball valve drain allows for quick and thorough tank draining
  • One quick coupler to easily support 1 nailer
  • Amperes: 2.60 A