Series 600 Trucks, Harper Trucks
Supplier: Safety & Industrial Supplies
338-60072-64 338-650A50-51 338-60068-77 338-65029-51 338-65050-51
300008-179EA 1308.23 USD
300008-179 300068-616 300068-610 300068-612 300068-614
Series 600 Trucks, Harper Trucks
Carts
Available in two and four wheel models and is designed to handle heavy CRYOGENIC LIQUID GAS CYLINDERS.
  • This larger sized frame and repositioned front wheels which allow secure handling without excessive force
  • The 650A50-51 is designed to handle larger 20" -26" diameter liquid gas cylinders
  • This model equipped with the QUAD Hook will handle a wide variety of large gas cylinders
  • Newly designed single hook adjusts from 50" to 60" in height which makes it more versatile for handling a variety of cylinder types
  • All available with pneumatic or mold on rubber
More Product Information