Industrial Series Carts, Saf-T-Cart
Supplier: Saf-T-Cart
339-LCT-12-6 339-730-12F
300083-672EA 1629.34 USD
300083-672 300083-626
Industrial Series Carts, Saf-T-Cart
Carts
The 730-12 and the 730-12F are multi-purpose carts designed specifically for the welding distributor; These carts will accommodate (1) liquid cylinder and (2) hi-pressure cylinders. The 730-12 has a fixed rear caster where the 730-12F has a folding rear caster for easy storage on a route truck.
The LCT-12-6 is a new design with spring load hanger; This cart is the same width as the cylinder for easy maneuverability through doorways and truck isles; It has 12" pneumatic front wheels and 6" rubber coated steel casters in the rear