In-Sight® Omni® Roll Towel Dispensers, Kimberly-Clark
Supplier: Kimberly-Clark
In-Sight® Omni®
412-09746
300070-482EA 122.39 USD
300070-482
In-Sight® Omni® Roll Towel Dispensers, Kimberly-Clark
Wipe Dispensers
Translucent, smoked plastic design allows you to see inside so you know exactly when a refill is needed without having to open the dispenser.
  • Easy to load
  • Cover protects towels
  • Serrated edge for easy tearing
  • When installed properly, this dispenser meets the ADA Standards for Accessible Design