Super Junior Vises, Wilton®, ORS Nasco
Supplier: ORS Nasco
Wilton®
825-63247
300034-880EA 249.37 USD
300034-880
Super Junior Vises, Wilton®, ORS Nasco
Vises
These are portable vises, the top jaws are grooved and replaceable.
  • Spindle is fully enclosed
  • Double lockdowns ensure stability
  • Can rotate 45° left or right on base
  • Locking type: threaded
  • Base type: Clamp-On
  • Jaw material: Steel