Torqueplus™ 12-Point Combination Wrenches - Satin Finish, Proto®, ORS Nasco
Supplier: Safety & Industrial Supplies
577-1216ASD 577-1222ASD 577-1228ASD 577-1248 577-1252 577-1264 577-1226ASD 577-1254 577-1256 577-1232ASD 577-1218ASD 577-1234ASD 577-1236ASD 577-1242 577-1266 577-1270 577-1210A 577-1212ASD 577-1214ASD 577-1220ASD 577-1224ASD 577-1230ASD 577-1240ASD 577-1244 577-1246 577-1208A 577-1258 577-1260 577-1268 577-1272 577-1276 577-1280
300002-716EA 33.68 USD
300002-716 300002-722 300002-726 300002-748 300002-750 300002-752 300023-752 300023-796 300023-800 300002-732 300002-718 300002-736 300002-738 300002-742 300023-814 300023-818 300002-706 300002-710 300002-714 300002-720 300002-724 300002-728 300002-740 300002-744 300002-746 300023-566 300023-804 300023-808 300023-816 300023-828 300023-830 300023-832
Torqueplus™ 12-Point Combination Wrenches - Satin Finish, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock