Reversible Ratcheting Box Wrenches, Proto®
Supplier: Proto®
577-1193LO 577-1194LO 577-1191LO
300023-498EA 53.78 USD
300023-498 300023-504 300023-492
Reversible Ratcheting Box Wrenches, Proto®
Wrenches
12 point box end allows for fine indexing, helping improve speed and access.
  • Built-in retaining ring in each ratchet wheel adds speed and convenience to great accessibility
  • Wrench self-aligns on the fastener which helps save time
  • Fits 4, 6 and 12 point fasteners