Metric Angle Open End Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
577-3109M 577-3115M 577-3119M 577-3117M 577-3116M 577-3112M 577-3114M 577-3118M 577-3113M 577-3111M 577-3110M
300024-558EA 46.11 USD
300024-558 300024-548 300024-554 300024-566 300024-544 300024-552 300024-564 300024-542 300024-550 300024-540 300024-560
Metric Angle Open End Wrenches, Proto®, ORS Nasco
Wrenches
For greater access in tight work areas.
  • 4 different turning angles
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Increased torque applied to fastener with fewer rounded edges
  • Unique arc design increases surface contact